Arsine generator systems
供应商: Corning Life Sciences
PYREX®
CORN3900EA 0 CNY
CORN3900 CORN7721-13
Arsine generator systems
消解系统 微量元素检测装置
This PYREX®, borosilicate glass arsine generator is designed for arsenic determination by the silver diethyldithiocarbamate photometric method. The 125 ml flask has a 24/40 inner standard taper joint and ball. Socket joint between the absorber and scrubber is a 12/2 spherical joint. Complete unit includes a clamp.