Centrefold film, fire retardant
供应商: Fisher Container
FISH2N600720FEA 5475 CNY
FISH2N600720F
Centrefold film, fire retardant
密封件 密封膜
Each roll of cleanroom film features a 182,9 cm centerfold and 30,5 m of film per roll.
Weight: 15,9 kg
Packaging: Film is packaged in a poly-lined carton.