Vertical electrophoresis systems, Mini-PROTEAN® Tetra cell
供应商: Bio-Rad
BRAD165-8004EA 0 CNY
BRAD165-8004
Vertical electrophoresis systems, Mini-PROTEAN® Tetra cell
电泳系统 垂直电泳系统
Mini-PROTEAN® Tetra vertical electrophoresis cell is easy to use and is ideal for protein electrophoresis with Bio-Rad precast polyacrylamide mini gels. It runs one to four Mini-PROTEAN® precast gels, Ready Gel® precast gels or handcast polyacrylamide gels.
订购须知 To run one to four gels, 4-gel cell or a 2-gel cell and the optional Mini-PROTEAN Tetra companion running module must be ordered separately.
Delivery information: Tetra cell includes tank, lid with power cables, electrode assembly, mini cell buffer dam, Mini-PROTEAN® Tetra companion running module.