Safety spectacles, Uvex Ultra-spec®
供应商: Honeywell Safety
uvex ultra-spec®
DALLS0300EA 0 CNY
DALLS0300
Safety spectacles, Uvex Ultra-spec®
眼镜
Economical safety glasses feature a 100% polycarbonate wrap around unilens design.
  • Clear lenses and frames
  • Adjustable temple length
  • Side shields add extra protection without compromising visibility
  • Ultra-dura® and Uvextreme® lens coating provides superior scratch and chemical resistance
Weight: 44 g