Isolator mitten, latex, AlphaTec® NRL 55-113
供应商: Ansell
AlphaTec®
ANSE55-113/10EA 0 CNY
ANSE55-113/10
Isolator mitten, latex, AlphaTec® NRL 55-113
手套
These hand-specific gauntlets are made ​​of natural rubber latex without an inner lining. They provide protection against chemical hazards as well as against abrasions and tears. For use with most available cleanroom glove boxes.
  • Highly resistant to most acids, alkalis, alcohols, aldehydes and ketones
  • Mitten shape provides effective protection for major risk environments
  • Medium thickness increases dexterity
  • Beaded cuffs for easier donning
  • Smooth finish
Ideal for use in chemical and medical research, biotechnologies, pharmaceutical industries and chemical/biological product handling.
Certifications: EN 388 (2020X), EN ISO 374-1 Type A (KLNOPT), EN ISO 374-5 VIRUS; Cat. III; AQL < 0,65