Spill kit, Vital 1®
供应商: North by Honeywell
Vital 1®
NORH127004EA 221 CNY
NORH127004
Spill kit, Vital 1®
泄露控制装置
Spill cleanup kit includes absorbent beads, gloves, red biohazard bags, germicidal surface towel, antiseptic towelette, shovel and absorbent towel. Flat sleeve-type box stacks neatly for storage.