Combination electrode, ORP, sensION™
供应商: Hach
sensION™
HACH5193700EA 3100 CNY
HACH5193700
Combination electrode, ORP, sensION™
电极 pH电极
This combination ORP/Redox electrode with platinum wire sensor features a replaceable reference electrolyte gel cartridge and an integrated temperature sensor. The electrode has a 5-pin connector.
  • Reference junction is resistant to clogging and easy to clean: Perfect for dirtier, more difficult samples where redox measurements are commonly made
  • Replaceable gel reference electrolyte cartridge is a fast and easy maintenance solution that maximise the life of the electrode
  • Rugged and durable: Designed to withstand the rigors of outdoor testing and frequent use
Compatible with Hach sensION systems.
Delivery information: Electrode is supplied with two replacement electrolyte gel cartridges and user manual.