This multi-parameter pH and ion-meter is perfect for research laboratories. It provides two measuring channels for pH/ORP/redox/ISE and option for two magnetic stirrers.
- Up to four ion selective calibration curves per channel; calibration with 2 to 5 standards
- Several calibration modes: with automatic recognition of 3 buffer groups, at a determined pH value, etc.
- Automatic buffer recognition: technical DIN 19267, DIN 19266 and up to 3 buffers chosen by the user
- Data logger storage of up to 330 measurements per channel
- RS232 C connectibility for printer or PC and connection for external PC, keyboard or barcode reader
Delivery information: Supplied with integrated magnetic stirrer and probe holder, power supply with interchangeable plugs, pH4, pH7 and pH10 calibration standard solutions (125 ml), KCl electrolyte solution (50 ml), 3 calibration beakers and printed documentation.