Multi-element calibration standard in dil. nitric acid for ICP-MS
供应商: VWR Chemicals
VWRC85026.180EA 8064 CNY
VWRC85026.180
Multi-element calibration standard in dil. nitric acid for ICP-MS
标准品 Cationic Standards Multi-Element Standards
30 Elements : 100 mg/L As, B, Be, Fe, Se, Zn ; 10 mg/L de Ba, Bi, Cd, Co, Cr, Cu, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Rb, Sr, Te, Ti, U, V ; 1000 mg/L Ca (HNO3 2%)