Retort clamp
供应商: R & L Enterprises
RLAAZ0104EA 0 CNY
RLAAZ0104
Retort clamp
夹具 通用夹具
Alloy die-cast, aluminium shaft. This clamp accepts articles up to 85 mm diameter. Actuated by nickel-plated brass pummels and blue plastic headed nickel-plated steel twin screw.
  • Fitted with an aluminium shaft
  • Four interlocking jaws are cork lined
This clamp accepts articles up to 85 mm diameter. Actuated by nickel-plated brass pummels and blue plastic headed nickel-plated steel twin screw.
  • Fitted with an aluminium shaft
  • Four interlocking jaws are cork lined