Small orifice cleaner, SMORC®, SP Bel-Art
供应商: Bel-Art
SMORC®
BELAF381250000EA 1822 CNY
BELAF381250000 BELAH381260000
Small orifice cleaner, SMORC®, SP Bel-Art
工具套件
Ideal for cleaning clogged nozzles, spray bottles, tubing and more.
A pin vise securely holds the stainless steel cleaning wire in place. A 22 mm Ø ring magnet on the handle is useful for picking up wires and for storing the unit securely on metal surfaces.
Delivery information: Supplied with wire assortment.