Multi-channel pipettes, variable volume, Cobra®
供应商: Biotix
BITXBTX-8X-300EA 7483 CNY
BITXBTX-12X-200 BITXBTX-12X-300 BITXBTX-8X-20 BITXBTX-8X-200 BITXBTX-8X-10 BITXBTX-12X-20 BITXBTX-8X-300 BITXBTX-12X-10
Multi-channel pipettes, variable volume, Cobra®
This lightweight and durable pipettor provides high accuracy and precision. It features a large arched finger hook design to provide all day comfort and reduce tension for all hand sizes. A higher hook means your thumb is much closer to the push button, thus reducing overall pipetting forces.
  • Soft spring system for extremely low pipetting forces; 360° rotating manifold to minimise hand and wrist movements
  • Adjustable tip-ejector button provides the best fit and ejection for right or left hand use for any hand size
  • Controlled volume setting works to prevent accidental volume changes
  • Stainless steel piston - better accuracy and precision over time for long-lasting results
  • Easy on site calibration (calibration key included)
  • Works with most tips on the market (tested for perfect fit with Biotix manual tips)
  • Fully autoclavable