Inline equilibrium cell, SP Bel-Art
供应商: Bel-Art
Scienceware®
BELAH403170000EA 6100 CNY
BELAH403170000
Inline equilibrium cell, SP Bel-Art
微孔板 细胞培养板
Clear acrylic construction with seven stainless steel screws and wing nuts for clamping cells and dialysis membrane. The cavity plug screws are Teflon® TFE coated stainless steel.
  • Each half cavity has a top opening for introduction or withdrawal of solutions and holds 1 ml in each half cell