Utility cart
供应商: Labconco
LBCP8007000EA 25313 CNY
LBCP8007000
Utility cart
手推车
These utility carts are compact, mobile work station for laboratory equipment. The upper open shelf transports and supports small electrical equipment. Has a lower 5,1 cm deep epoxy-coated steel with removable watertight pan transports wet or soiled glassware and other labware. Power strip with 115 volts, 60 Hz, 12 amps capacity, has 6 electrical outlets, cord bracket and a 12-foot, 3-wire cord with plug.
  • Welded steel frames, 18-gauge of 1 inch square tubing
  • Painted with tough, corrosion-resistant, dry powder epoxy
  • Two corrosion-resistant, one-piece, vacuum-formed ABS plastic drawers with 10 cm diameter casters, two with toe locks