Chemical cart
供应商: Labconco
LBCP8020000EA 17131.1 CNY
LBCP8020000
Chemical cart
手推车
Chemical cart, has two open shelves with rigid PVC extrusions on two sides to prevent containers from sliding off. The lower shelf also has rigid plastic guard rails on both ends.
  • With 4" Ø casters, two with toe locks
  • Ready-to-roll, full factory assembly
  • Ergonomic, extruded rubber handle grips
  • Durable glacier white, epoxy-coated 18 gauge welded tubular steel frame