Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 2C11]
供应商: Abnova
ABNOH00008878-M01EA 0 CNY
ABNOH00008878-M01
Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 2C11]
抗体
Type: Primary
Antigen: SQSTM1
Clonality: Monoclonal
Clone: 2C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse