Anti-DDR1 Mouse monoclonal antibody unconjugated

供应商: BIORBYT
BIRBORB69032-100EA 0 CNY
BIRBORB69032-100
Anti-DDR1 Mouse monoclonal antibody unconjugated
抗体
Western Blot: 1:500-1:2000; ELISA: 1:10000

Type: Primary
Antigen: DDR1 (discoidin domain receptor tyrosine kinase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR