Anti-Pellino 1 Rabbit Polyclonal Antibody

供应商: Abcam
ABCAAB13812-200EA 3500 CNY
ABCAAB13812-200
Anti-Pellino 1 Rabbit Polyclonal Antibody
抗体
Type: Primary
Antigen: Pellino 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR