Anti-COX IV Mouse Monoclonal Antibody [clone: 20E8C12]
供应商: Abcam
ABCAAB14744-100EA 0 CNY
ABCAAB14744-100
Anti-COX IV Mouse Monoclonal Antibody [clone: 20E8C12]
抗体
Type: Primary
Antigen: COX IV
Clonality: monoclonal
Clone: 20E8C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
现在订购