Anti-CDK9 Rabbit polyclonal antibody unconjugated

供应商: Abcam
ABCAAB39098-200EA 0 CNY
ABCAAB39098-200
Anti-CDK9 Rabbit polyclonal antibody unconjugated
抗体
Type: Primary
Antigen: CDK9 (cyclin-dependent kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR