Anti-NGF Rabbit Monoclonal Antibody [clone: EP1320Y]

供应商: Abcam
ABCAAB52918-100EA 0 CNY
ABCAAB52918-100
Anti-NGF Rabbit Monoclonal Antibody [clone: EP1320Y]
抗体
Type: Primary
Antigen: NGF (nerve growth factor (beta polypeptide))
Clonality: monoclonal
Clone: EP1320Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR