Anti-MTHFD2 Mouse Monoclonal Antibody

供应商: Abcam
ABCAAB56772-100EA 0 CNY
ABCAAB56772-100
Anti-MTHFD2 Mouse Monoclonal Antibody
抗体
Type: Primary
Antigen: MTHFD2 (methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR