Anti-ELOVL6 Rabbit polyclonal antibody unconjugated

供应商: Abcam
ABCAAB69857-100EA 0 CNY
ABCAAB69857-100
Anti-ELOVL6 Rabbit polyclonal antibody unconjugated
抗体
Type: Primary
Antigen: ELOVL6 (ELOVL fatty acid elongase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR