Anti-IgG Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
供应商: Abcam
ABCAAB106718-100EA 0 CNY
ABCAAB106718-100
Anti-IgG Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
抗体
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
现在订购