Anti-Cytochrome C oxidase subunit II Mouse Monoclonal Antibody [clone: 12C4F12]
供应商: Abcam
ABCAAB110258-100EA 0 CNY
ABCAAB110258-100
Anti-Cytochrome C oxidase subunit II Mouse Monoclonal Antibody [clone: 12C4F12]
抗体
Type: Primary
Antigen: Cytochrome C oxidase subunit II
Clonality: monoclonal
Clone: 12C4F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
现在订购