Anti-IRAK4 Mouse Monoclonal Antibody [clone: 2H9]

供应商: Abcam
ABCAAB119942-100EA 0 CNY
ABCAAB119942-100
Anti-IRAK4 Mouse Monoclonal Antibody [clone: 2H9]
抗体
Type: Primary
Antigen: IRAK4 (interleukin-1 receptor-associated kinase 4)
Clonality: monoclonal
Clone: 2H9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR