Anti-CD27 Rabbit Monoclonal Antibody [clone: EPR8569]
供应商: Abcam
ABCAAB131254-100EA 0 CNY
ABCAAB131254-100
Anti-CD27 Rabbit Monoclonal Antibody [clone: EPR8569]
抗体
Type: Primary
Antigen: CD27 (CD27 molecule)
Clonality: monoclonal
Clone: EPR8569
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
现在订购