Anti-CD55 Rabbit Monoclonal Antibody

供应商: Abcam
ABCAAB133684-100EA 3650 CNY
ABCAAB133684-100
Anti-CD55 Rabbit Monoclonal Antibody
抗体
Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR