Anti-CD59 Rabbit Monoclonal Antibody [clone: EPR6425]

供应商: Abcam
ABCAAB133707-100EA 0 CNY
ABCAAB133707-100
Anti-CD59 Rabbit Monoclonal Antibody [clone: EPR6425]
抗体
Type: Primary
Antigen: CD59 (CD59 molecule, complement regulatory protein)
Clonality: monoclonal
Clone: EPR6425
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR