Anti-NCAM Rabbit Monoclonal Antibody

供应商: Abcam
ABCAAB137086-100EA 0 CNY
ABCAAB137086-100
Anti-NCAM Rabbit Monoclonal Antibody
抗体
Type: Primary
Antigen: NCAM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR