Anti-TLR4 Mouse Monoclonal Antibody [clone: 76B357.1]

供应商: Novus Biologicals
NOVUNB100-56566EA 0 CNY
NOVUNB100-56566
Anti-TLR4 Mouse Monoclonal Antibody [clone: 76B357.1]
抗体
The TLR4 Antibody (76B357.1) from Novus Biologicals is a mouse monoclonal antibody to TLR4. This antibody reacts with human, mouse, rat. The TLR4 Antibody (76B357.1) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: 76B357.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR