Anti-TLR9 Mouse Monoclonal Antibody [clone: 26C593.2]

供应商: Novus Biologicals
NOVUNBP2-24729EA 0 CNY
NOVUNBP2-24729
Anti-TLR9 Mouse Monoclonal Antibody [clone: 26C593.2]
抗体
Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: monoclonal
Clone: 26C593.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR