Anti-TLR5 Mouse Monoclonal Antibody [clone: 19D759.2]

供应商: Novus Biologicals
NOVUNBP2-24787EA 0 CNY
NOVUNBP2-24787
Anti-TLR5 Mouse Monoclonal Antibody [clone: 19D759.2]
抗体
Type: Primary
Antigen: TLR5 (toll-like receptor 5)
Clonality: monoclonal
Clone: 19D759.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR