Anti-TLR8 Mouse Monoclonal Antibody [clone: 44C143]

供应商: Novus Biologicals
NOVUNBP2-24917EA 0 CNY
NOVUNBP2-24917
Anti-TLR8 Mouse Monoclonal Antibody [clone: 44C143]
抗体
Type: Primary
Antigen: TLR8 (toll-like receptor 8)
Clonality: monoclonal
Clone: 44C143
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR