Anti-Nitric Oxide Synthase-Inducible Rabbit Monoclonal Antibody [clone: SP126]

供应商: Thermo Scientific
PIEAMA516422EA 0 CNY
PIEAMA516422
Anti-Nitric Oxide Synthase-Inducible Rabbit Monoclonal Antibody [clone: SP126]
抗体
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is lung tissue. Inducible NOS (iNOS) or macrophage NOS (mNOS) is calcium/calmodulin independent and is expressed in activated macrophages and stimulated glial cells. NOS oxidizes a guanidine nitrogen of arginine, releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation, neurotransmission, anti-tumor and anti-pathogenic activities.

Type: Primary
Antigen: Nitric Oxide Synthase-Inducible
Clonality: Monoclonal
Clone: SP126
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR