Anti-Fluorescein Rabbit polyclonal antibody unconjugated

供应商: Thermo Fisher Scientific
PIEAPA128347EA 0 CNY
PIEAPA128347
Anti-Fluorescein Rabbit polyclonal antibody unconjugated
抗体
Type: Primary
Antigen: Fluorescein (fluorescein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR