Anti-NRROS Rabbit Polyclonal Antibody

供应商: Thermo Scientific
PIEAPA523822EA 0 CNY
PIEAPA523822
Anti-NRROS Rabbit Polyclonal Antibody
抗体
Type: Primary
Antigen: NRROS (negative regulator of reactive oxygen species)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR