Anti-IgY Goat Polyclonal Antibody (DyLight® 350)

供应商: Thermo Fisher Scientific
PIEASA510069EA 0 CNY
PIEASA510069
Anti-IgY Goat Polyclonal Antibody (DyLight® 350)
抗体
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR