Anti-IgG Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

供应商: Cytiva
GEHENA933-1MLEA 0 CNY
GEHENA933-1ML
Anti-IgG Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
抗体
Highly species specific HRP-conjugated antibodies optimized for use with Amersham ECL Western Blotting Detection Reagents.

  • Optimized for use with our range of Amersham ECL Western blotting detection reagents
  • Soluble in cold water and hot water

The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2 °C to 8 °C.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR