Anti-BrdU Mouse Monoclonal Antibody

供应商: Cytiva
GEHERPN202EA 0 CNY
GEHERPN202
Anti-BrdU Mouse Monoclonal Antibody
抗体
Anti-Bromodeoxyuridine (Anti-BRdU) antibody is supplied in the same formulation as the monoclonal antibody to BrdU in the Cell Proliferation Kit. The antibody/nuclease mixture is compatible with detection systems that incorporate fluorescently labelled antibodies and biotin-streptavidin detection reagents.

  • No need for chemical denaturation
  • Detection while maintaining cell morphology
  • Double immunocytochemical staining studies can be perfromed

Used in conjunction with RPN20, the reagent allows extended use of the kit where additional amounts of labeling reagent are required, such as in vivo studies.

The antibody contains a nuclease to produce single stranded DNA without the need for chemical denaturation. Cellular morphology is preserved allowing multiple labeling experiments to be carried out. The high-contrast stain produced has sensitivity comparable to autoradiographic methods.

Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Chemical
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR