Cleanroom goggles, V80 REVOLUTION*, JACKSON SAFETY*
供应商: Kimberly-Clark
JACKSON SAFETY*
KIMB14399EA 0 CNY
KIMB14399
Cleanroom goggles, V80 REVOLUTION*, JACKSON SAFETY*
护目镜
These cleanroom goggles can be worn over most prescription eyeglasses.
  • CROSS VENT* technology maintains airflow and minimises moisture build-up
  • VISICLEAR* anti-fog and anti-scratch lens coating
  • Expanded visual field
Certifications: Meets ANSI Z87.1+ impact standard