Rockers, compact, digital
供应商: Thermo Scientific
BARN88880022EA 8550 CNY
BARN88880022 BARN88880020
Rockers, compact, digital
This digital rocker ensures gentle sample agitation with compact, adjustable angle and digital display unit. Designed for time-dependent and low-dependent sensitive applications such as ELISA, protein synthesis, staining/de-staining gels, and antibody staining.
  • PID control ensures consistent and smooth rocking motion
  • Ideal for placing in incubators (non-CO₂), chambers and refrigerators (up to 60 °C)
Speed acuracy (at 100 min⁻¹): ±5 min⁻¹
Delivery information: Supplied with dimpled mat (tray, silicone pad, two securing bands). Flat sticky mat tray or deep ledge sticky mat tray for plates, dishes and bags should be ordered separately.