Heat resistant autoclave gloves, Jomac® KELKLAVE
供应商: Wells Lamont
Jomac®
WELL422-11EA 0 CNY
WELL422-11 WELL422-5
Heat resistant autoclave gloves, Jomac® KELKLAVE
手套
These autoclave gloves made from white loop-out terry cloth provide protection against temperatures up to 121 °C. Ideal for use when removing parts from autoclaves.
  • Available with short or long gauntlet cuff
  • Clute cut
  • Machine washable