Heat resistant gloves, Jomac®
供应商: Wells Lamont
Jomac®
WELL9001EA 0 CNY
WELL9001
Heat resistant gloves, Jomac®
手套
These seamless medium-weight Kevlar/cotton blend terry cloth gloves provide protection against heat hazards and cuts.
  • Protects against temperatures up to 93 °C
  • Knit wrist
  • Loop-out
  • Ambidextrous
  • Machine washable