High vacuum valves, with PTFE plug, HI-VAC®, KIMBLE®
供应商: DWK Life Sciences
KIML826620-0004EA 0 CNY
KIML826620-0004
High vacuum valves, with PTFE plug, HI-VAC®, KIMBLE®
阀门 真空阀
These high vacuum valves with PTFE plug are made from borosilicate glass.
  • Seal design reduces outgassing
  • Designed with a precision external thread for fine control
  • With low hold up
  • Non-rotating PTFE shaft
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.