Glass cutter, diamond, Synthware
供应商: Kemtech America
KEMTDC18001EA 0 CNY
KEMTDC18001
Glass cutter, diamond, Synthware
刀具
The glass cutter features a rack on shank for breaking off single or double strength glass. Extra-large natural diamond is used for durability.