Human pulmonary artery endothelial cells (HPAEC)
供应商: PromoCell
POMOC-12241EA 0 CNY
POMOC-12241 POMOC-14024
Human pulmonary artery endothelial cells (HPAEC)
细胞
Primary Human Pulmonary Artery Endothelial Cells (HPAEC) are isolated from human pulmonary arteries (the main pulmonary artery, left and right branches)
  • Von Willebrand factor (vWF) positive
  • CD31 positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
Since the pulmonary arteries carry blood from the heart to the lungs, they are the only arteries (other than umbilical arteries) that carry deoxygenated blood.
订购须知 Human Pulmonary Artery Endothelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.