Filtering beakers, DURAN®
供应商: DWK Life Sciences
SCOT211339101EA 7773 CNY
SCOT211339101
Filtering beakers, DURAN®
烧杯
DURAN®, borosilicate glass 3.3, clear.
  • Increased wall thickness for better mechanical properties
  • Reduced thermal shock resistance
  • Limited application for heating
Thick walled, without spout.