Mouse IgG1 kappa Isotype Control [clone: MG1-45]

供应商: BioLegend
BLEG401401EA 0 CNY
BLEG401402 BLEG401401
Mouse IgG1 kappa Isotype Control [clone: MG1-45]
抗体
Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MG1-45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR