Anti-PDCD1LG2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 24F.10C12]

供应商: BioLegend
BLEG329606EA 0 CNY
BLEG329606 BLEG329605
Anti-PDCD1LG2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 24F.10C12]
抗体
Type: Primary
Antigen: PDCD1LG2
Clonality: Monoclonal
Clone: 24F.10C12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR